Product Name
Nuarimol PESTANAL®, analytical standard
Product Number
31116
Product Brand
SIAL
CAS Number
63284-71-9
Molecular Weight
314.74
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Light Yellow to Light Beige |
APPEARANCE (FORM) | Powder or Crystals |
PURITY (HPLC AREA %) | ≥ 98.0 % |
MELTING POINT | 123 - 128 C |
WATER | ≤ 1.0 % |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 60 MONTHS |