Product Name
Phenethylamine ≥99%
Product Number
241008
Product Brand
ALDRICH
CAS Number
64-04-0
Molecular Weight
121.18
Test | Specification |
---|---|
Appearance (Color) | Colorless |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Purity (GC) | ≥99.0 % |