Product Name
(+)-3-(Trifluoroacetyl)camphor 98%
Product Number
189006
Product Brand
ALDRICH
CAS Number
51800-98-7
Molecular Weight
248.24
Test | Specification |
---|---|
Appearance (Color) | Colorless |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Optical Rotation | 142 - 154 deg |
c = 2.3 %; CH2CL2 | |
Purity (GC) | ≥97.5 % |