Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~110 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(E-A-SP-G-A)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer213)

Gene Information

human ... AR(367)

Immunogen

Peptide sequence around phosphorylation site of serine 213 (E-A-S(p)-G-A), according to the protein AR.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The androgen receptor gene is more than 90 kb long and codes for a protein that has 3 major functional domains: theN-terminal domain, DNA-binding domain, and androgen-binding domain. The protein functions as a steroid-hormoneactivated transcription factor. Upon binding the hormone ligand, the receptor dissociates from accessory proteins,translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. This genecontains 2 polymorphic trinucleotide repeat segments that encode polyglutamine and polyglycine tracts in theN-terminal transactivation domain of its protein. Expansion of the polyglutamine tract causes spinal bulbar muscularatrophy (Kennedy disease). Mutations in this gene are also associated with complete androgen insensitivity (CAIS). Twoalternatively spliced variants encoding distinct isoforms have been described.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service