SAB1410046
Anti-USP3 antibody produced in mouse
purified immunoglobulin, buffered aqueous solution
Synonym(s):
MGC129878, MGC129879, SIH003, UBP
Select a Size
Select a Size
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 58.9 kDa
species reactivity
human
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 1 μg/mL
Immunogen
Sequence
MECPHLSSSVCIAPDSAKFPNGSPSSWCCSVCRSNKSPWVCLTCSSVHCGRYVNGHAKKHYEDAQVPLTNHKKSEKQDKVQHTVCMDCSSYSTYCYRCDDFVVNDTKLGLVQKVREHLQNLENSAFTADRHKKRKLLENSTLNSKLLKVNGSTTAICATGLRNLGNTCFMNAILQSLSNIEQFCCYFKELPAVELRNGKTAGRRTYHTRSQGDNNVSLVEEFRKTLCALWQGSQTAFSPESLFYVVWKIMPNFRGYQQQDAHEFMRYLLDHLHLELQGGFNGVSRSAILQENSTLSASNKCCINGASTVVTAIFGGILQNEVNCLICGTESRKFDPFLDLSLDIPSQFRSKRSKNQENGPVCSLRDCLRSFTDLEELDETELYMCHKCKKKQKSTKKFWIQKLPKVLCLHLKRFHWTAYLRNKVDTYVEFPLRGLDMKCYLLEPENSGPESCLYDLAAVVVHHGSGVGSGHYTAYATHEGRWFHFNDSTVTLTDEETVVKAKAYILFYVEHQAKAGSDKL
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service