biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
western blot: 1 μg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PRKCQ(5588)
General description
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role. The protein encoded by this gene is one of the PKC family members. It is a calcium-independent and phospholipid-dependent protein kinase. This kinase is important for T-cell activation. It is required for the activation of the transcription factors NF-kappaB and AP-1, and may link the T cell receptor (TCR) signaling complex to the activation of the transcription factors. (provided by RefSeq)
Immunogen
PRKCQ (NP_006248.1, 1 a.a. ~ 706 a.a) full-length human protein.
Sequence
MSPFLRIGLSNFDCGSCQSCQGEAVNPYCAVLVKEYVESENGQMYIQKKPTMYPPWDSTFDAHINKGRVMQIIVKGKNVDLISETTVELYSLAERCRKNNGKTEIWLELKPQGRMLMNARYFLEMSDTKDMNEFETEGFFALHQRRGAIKQAKVHHVKCHEFTATFFPQPTFCSVCHEFVWGLNKQGYQCRQCNAAIHKKCIDKVIAKCTGSAINSRETMFHKERFKIDMPHRFKVYNYKSPTFCEHCGTLLWGLARQGLKCDACGMNVHHRCQTKVANLCGINQKLMAEALAMIESTQQARCLRDTEQIFREGPVEIGLPCSIKNEARPPCLPTPGKREPQGISWESPLDEVDKMCHLPEPELNKERPSLQIKLKIEDFILHKMLGKGSFGKVFLAEFKKTNQFFAIKALKKDVVLMDDDVECTMVEKRVLSLAWEHPFLTHMFCTFQTKENLFFVMEYLNGGDLMYHIQSCHKFDLSRATFYAAEIILGLQFLHSKGIVYRDLKLDNILLDKDGHIKIADFGMCKENMLGDAKTNTFCGTPDYIAPEILLGQKYNHSVDWWSFGVLLYEMLIGQSPFHGQDEEELFHSIRMDNPFYPRWLEKEAKDLLVKLFVREPEKRLGVRGDIRQHPLFREINWEELERKEIDPPFRPKVKSPFDCSNFDKEFLNEKPRLSFADRALINSMDQNMFRNFSFMNPGMERLIS
Sequence
MSPFLRIGLSNFDCGSCQSCQGEAVNPYCAVLVKEYVESENGQMYIQKKPTMYPPWDSTFDAHINKGRVMQIIVKGKNVDLISETTVELYSLAERCRKNNGKTEIWLELKPQGRMLMNARYFLEMSDTKDMNEFETEGFFALHQRRGAIKQAKVHHVKCHEFTATFFPQPTFCSVCHEFVWGLNKQGYQCRQCNAAIHKKCIDKVIAKCTGSAINSRETMFHKERFKIDMPHRFKVYNYKSPTFCEHCGTLLWGLARQGLKCDACGMNVHHRCQTKVANLCGINQKLMAEALAMIESTQQARCLRDTEQIFREGPVEIGLPCSIKNEARPPCLPTPGKREPQGISWESPLDEVDKMCHLPEPELNKERPSLQIKLKIEDFILHKMLGKGSFGKVFLAEFKKTNQFFAIKALKKDVVLMDDDVECTMVEKRVLSLAWEHPFLTHMFCTFQTKENLFFVMEYLNGGDLMYHIQSCHKFDLSRATFYAAEIILGLQFLHSKGIVYRDLKLDNILLDKDGHIKIADFGMCKENMLGDAKTNTFCGTPDYIAPEILLGQKYNHSVDWWSFGVLLYEMLIGQSPFHGQDEEELFHSIRMDNPFYPRWLEKEAKDLLVKLFVREPEKRLGVRGDIRQHPLFREINWEELERKEIDPPFRPKVKSPFDCSNFDKEFLNEKPRLSFADRALINSMDQNMFRNFSFMNPGMERLIS
Physical form
Solution in phosphate buffered saline, pH 7.4
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service