Skip to Content
Merck
All Photos(2)

Key Documents

OM-170047X

OMICRON™

OMICRON™ Nylon Membrane Filters

Nylon, 0.45 Micron, 47mm (100/Pk)

Synonym(s):

Nylon Membrane, Filters, Membrane filters

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item


PDF Icon
Shipped & Sold By Omicron Scientific
A Trusted Marketplace Seller
PDF Icon
Have a question?

manufacturer/tradename

Omicron Scientific

General description

OMICRON™ Nylon Membrane Filter

OMICRON™ Nylon membrane filters are ideal for use in general filtration, medical assays, protein binding and can stand up to solvent solution.

Technical Specifications
  • Color: White
  • Surface: Plain
  • Wettability: Hydrophilic
  • Extractable: Very Low (Within limit as specified in USP XXIV)
  • Heat Stability: Maintain integrity for 5 cycles of autoclave at 121°C for 30 minutes.
  • Operating Temp.: 75°C
Application (Sterile)
  • Filtration of microbiological media, sera and other additives.
  • Ideal for HPLC Solvent.
  • Sterilizing filtration of pharmaceuticals aqueous solution and bioassay.
  • Filtraion of wide range of injectable solution.
  • Ideal for organic solvent.
Application (Non-Sterile)
  • Ideal for HPLC Solvents.
  • Mobile phase separation and solvent filtration.

(Closely similar to 7404-004)

Legal Information

OMICRON™ is a trademark of Omicron Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library