Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCR028RF

Sigma-Aldrich

Mesenchymal Stem Cell Osteogenesis Kit (2-8C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service