Skip to Content
Merck
All Photos(1)

Key Documents

MABN122

Sigma-Aldrich

Anti-Robo2 Antibody, clone 4C6.1

clone 4C6.1, from mouse

Synonym(s):

Roundabout homolog 2

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4C6.1, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

General description

Roundabout homolog 2 (Robo2) is a receptor for SLIT2, and likely SLIT1, which are thought to help guide cellular migration, including axonal navigation of the neural tube and the projection of axons during neuronal development. Vesicoureteral reflux type 2 (VUR2) has been attributed to defects in Robo2.

Specificity

This antibody recognizes the cytoplasmic domain of Robo2

Immunogen

Epitope: Cytoplasmic domain
GST-tagged recombinant protein corresponding to the cytoplasmic domain of human Robo2.

Application

Detect Roundabout homolog 2 using this mouse monoclonal antibody, Anti-Robo2 Antibody, clone 4C6.1 validated for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Robo2 in rat brain (hypothalamus) and human brain tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in mouse neuronal stem cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Robo2 in 200 µg of mouse neuronal stem cell lysate.

Target description

~ 170 kDa observed. The calculated molecular weight is 150 kDa, however Robo2 has been shown as a ~170 kDa band in western blots due to high glycosylation. (Fricke, C., et al. (2001). Science. 292:(5516):507-510).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse neuronal stem cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service