Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6F3.1, monoclonal
species reactivity
rat
species reactivity (predicted by homology)
human (based on 100% sequence homology)
technique(s)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... RET(5979)
General description
Proto-oncogene tyrosine-protein kinase receptor Ret (EC 2.7.10.1; UniProt P07949; also known as Cadherin family member 12, Cadherin-related family member 16, Hydroxyaryl-protein kinase, Proto-oncogene c-Ret, Rearranged during transfection, Receptor tyrosine kinase, Ret proto-oncogene, RET receptor tyrosine kinase, RET transforming sequence) is encoded by the RET (also known as CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET-ELE1, RET51) gene (Gene ID 5979) in human. Ret is a receptor tyrosine kinase that mediates the activation of several signaling pathways including the PLC gamma, Ras, JNK and inositol phosphate pathways. Ret mutations have been shown to be causative in several diseases, including Hirschsprung′s disease (HD), papillary thyroid carcinoma, multiple endocrine neoplasia (MEN) 2A, MEN 2B, and familial medullary thyroid carcinoma. Ret is initially produced with a signal peptide sequence (a.a. 1-28), the removal of which yields the mature Ret (a.a. 20-1114). Ret can be posttranslationally cleaved into the extracellular cell-membrane-anchored RET cadherin 120 kDa fragment (a.a. 29-707) and the soluble RET kinase fragment (a.a. 708-1017).
Specificity
Expected to react with both the RET51 and the RET9 isoforms, as well as the extracellular cell-membrane anchored RET cadherin 120 kDa fragment (a.a. 29-707).
Immunogen
Epitope: Extracellular domain
GST-tagged recombinant protein corresponding to the extracellular domain of human c-Ret.
Application
Research Category
Apoptosis & Cancer
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Apoptosis - Additional
This Anti-c-Ret Antibody, clone 6F3.1 is validated for use in Western Blotting for the detection of c-Ret.
Quality
Evaluated by Western Blotting in PC12 cell lysate.
Western Blotting Analysis: 1.0 µg/mL of this antibody detected c-Ret in 10 µg of PC12 cell lysate.
Western Blotting Analysis: 1.0 µg/mL of this antibody detected c-Ret in 10 µg of PC12 cell lysate.
Target description
~150 kDa observed. 124.3 kDa (RET51) and 119.8 kDa (RET9) calculated. Target band(s) may appear larger than the calculated molecular size(s) due to posttranslational glycosylation and/or phosphorylation. Uncharacterized band(s) may appear in some lysates.
Physical form
Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service