Skip to Content
MilliporeSigma
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Induced Pluripotent Stem Cells
Mouse Embryonic Stem Cells
Neural Stem Cells

Quality Level

technique(s)

cell based assay: suitable

detection method

colorimetric

General description

Extracellular matrix (ECM) proteins are produced intracellularly and are subsequently secreted into the surrounding cellular medium, actively regulating a diverse range of cell functions including cell adhesion, differentiation, proliferation, migration, invasion and survival. A primary utility of ECMs in in vitro culture is to promote cellular adhesion while maintaining cell viability and maximizing cell proliferation for downstream cell-based applications. In cases where optimal cell growth conditions are not well defined and critical adhesion protein requirements are unknown, identifying the ideal ECM protein(s) at functionally relevant concentration(s) can be a time-consuming and labor-intensive process. The 48-well, colorimetric ECM Cell Culture Optimization Array provides a multiparametric assay that allows researchers to: (1) quickly identify ECM(s) that best promote cell-type specific adhesion and (2) pinpoint the optimal ECM concentration required to promote maximum adhesion for a particular cell type.

How does the ECM Cell Culture Optimization Array work?
The foundation of our kit is a 48-well microtiter plate containing several commonly-used ECM proteins. Collagen I, laminin, fibronectin, and vitronectin are arrayed in triplicate with concentrations ranging from 0.1 μg/mL to 20 μg/mL. You simply culture your cells on the plate, and use the included reagents to block, stain, and analyze the adhesion capacity with a plate reader. We have extensively tested and optimized this kit on a variety of cell types including HEK293 cells, and human and rodent neural stem cells.

Application

The ECM Cell Culture Optimization Array is the first commercially-available tool of its kind to enable researchers to not only quickly identify the best ECM protein for their cell culture environment, but also pinpoint the concentration needed to achieve optimal cell growth conditions.

Packaging

1 plate, 48-wells

Components

  • ECM Gradient Array Plate: One 48-well plate • 30%BSA: (Part No. CS201638) One vial - 0.5mL • Cell Stain Solution: (Part No. 90144) One bottle - 20 mL. • Extraction Buffer: (Part No. 90145) One bottle - 20 mL • Assay Buffer: (Part No. 2003646) One bottle - 100 mL

Storage and Stability

The ECM Gradient Array Plate should be stored in the foil pouch at -70°C, and used within three months of the date of receipt. The remaining kit components should be stored at 2° to 8°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service