Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN259

Sigma-Aldrich

Anti-PANK2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Pantothenate kinase 2, mitochondrial, hPanK2, Pantothenic acid kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PANK2(80025)

General description

PANK2 (Pantothenate kinase 2) phosphorylates pantothenic acid (vitamin B(5) and regulates the overall rate of coenzyme A (CoA) biosynthesis. PANK2 thereby plays an important role in lipid metabolism and energy production. Mutation or knockdown of the PANK2 gene results in various disease states, including hypoglycemia, hyperketonemia, faulty metabolism, poor postnatal development, and childhood neurodegeneration.

Specificity

This antibody demonstrates specificity for isoform 1, but does not cross react with other isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to human PANK2.

Application

Detect the PANK2 protein using this Anti-PANK2 validated for use in WB.

Quality

Evaluated by Western Blot in human hippocampus tissue lysate.

Western Blot Analysis: 1.0 µg/mL of this antibody detected PANK2 in 10 µg of human hippocampus tissue lysate.

Target description

~62/37 kDa observed. Uncharacterized bands may appear at ~15 kDa, ~25 kDa, and ~260 kDa in some lysates. The calculated molecular weight is 62 kDa, however PANK2 has been shown as a ~40 kDa band in western blots (Kotzbauer, P. T., et al. (2005). The Journal of Neuroscience. 25(3):689-698).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service