MilliporeSigma
All Photos(7)
Key Documents
BAF371200125
SP Bel-Art® Spinbar® magnetic stir bar
polygon white PTFE-coated (no pivot ring), L × diam. 0.5 in. (12 mm) × 0.1875 in. (5 mm)
Synonym(s):
Spinbar® magnetic stir bar
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
material
polygon white PTFE-coated (no pivot ring)
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37120-0125
L × diam.
0.5 in. (12 mm) × 0.1875 in. (5 mm)
General description
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
High quality PTFE encapsulated magnetic stirring bars ensure efficient mixing with consistent results.
Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
Features and Benefits
  • Multi-faceted polygon shape with increased surface area creates added turbulence relative to similar sized, smooth cylindrical bars to provide thorough mixing
  • Absence of a pivot ring allow the Spinbar to stay close to the bottom of the vessel enabling “scraping” of solids
  • Optimal coupling with magnetic spindrive as entire length of stir bar is in close contact
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating; superior chemical compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)