HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 5 mM ammonium acetate, pH 4.1; [B] acetonitrile; (70:30, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
null