HPLC Analysis of FMOC-Norvaline Enantiomers on Astec® CHIROBIOTIC® T
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (60:40, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 280 nm
injection
0.5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC