GC Analysis of Patchouli Essential Oil on Equity-1
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1, 30 m x 0.25 mm I.D., 0.25 μm (28046-U)
oven
50 °C (0.5 min), then 5 °C/min to 300 °C (10 min)
inj. temp.
280 °C
detector
FID, 310 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL, 100:1 split
liner
4 mm I.D. cup injector
sample
Patchouli Oil, neat (W283800)
Description
Legal Information
null