Merck
  • Differential effects of type 2 diabetes on brain glycometabolism in rats: focus on glycogen and monocarboxylate transporter 2.
Differential effects of type 2 diabetes on brain glycometabolism in rats: focus on glycogen and monocarboxylate transporter 2.
The journal of physiological sciences : JPS (2016-12-18)
Takeru Shima, Subrina Jesmin, Takashi Matsui, Mariko Soya, Hideaki Soya
ABSTRACT
Astrocyte-neuron lactate shuttle (ANLS) is a pathway that supplies glycogen-derived lactate to active neurons via monocarboxylate transporter 2 (MCT2), and is important for maintaining brain functions. Our study revealed alterations of ANLS with hippocampal hyper-glycogen levels and downregulated MCT2 protein levels underlying hippocampal dysfunctions as a complication in type 2 diabetic (T2DM) animals. Since T2DM rats exhibit brain dysfunctions involving several brain regions, we examined whether there might also be T2DM effects on ANLS's disturbances in other brain loci. OLETF rats exhibited significantly higher glycogen levels in the hippocampus, hypothalamus, and cerebral cortex than did LETO rats. MCT2 protein levels in OLETF rats decreased significantly in the hippocampus and hypothalamus compared to their controls, but a significant correlation with glycogen levels was only observed in the hippocampus. This suggests that the hippocampus may be more vulnerable to T2DM compared to other brain regions in the context of ANLS disruption.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid