Merck
  • Inhibition of C1-Ten PTPase activity reduces insulin resistance through IRS-1 and AMPK pathways.
Inhibition of C1-Ten PTPase activity reduces insulin resistance through IRS-1 and AMPK pathways.
Scientific reports (2017-12-21)
Heeyoon Jeong, Ara Koh, Jiyoun Lee, Dohyun Park, Jung Ok Lee, Mi Nam Lee, Kyung-Jin Jo, Huynh Nguyen Khanh Tran, Eui Kim, Byung-Sun Min, Hyeon Soo Kim, Per-Olof Berggren, Sung Ho Ryu
ABSTRACT
Insulin resistance causes type 2 diabetes; therefore, increasing insulin sensitivity is a therapeutic approach against type 2 diabetes. Activating AMP-activated protein kinase (AMPK) is an effective approach for treating diabetes, and reduced insulin receptor substrate-1 (IRS-1) protein levels have been suggested as a molecular mechanism causing insulin resistance. Thus, dual targeting of AMPK and IRS-1 might provide an ideal way to treat diabetes. We found that 15,16-dihydrotanshinone I (DHTS), as a C1-Ten protein tyrosine phosphatase inhibitor, increased IRS-1 stability, improved glucose tolerance and reduced muscle atrophy. Identification of DHTS as a C1-Ten inhibitor revealed a new function of C1-Ten in AMPK inhibition, possibly through regulation of IRS-1. These findings suggest that C1-Ten inhibition by DHTS could provide a novel therapeutic strategy for insulin resistance-associated metabolic syndrome through dual targeting of IRS-1 and AMPK.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)