Merck
Sez6l2 regulates phosphorylation of ADD and neuritogenesis.
Biochemical and biophysical research communications (2017-10-17)
Hiroaki Yaguchi, Ichiro Yabe, Hidehisa Takahashi, Masashi Watanabe, Taichi Nomura, Takahiro Kano, Masaki Matsumoto, Keiichi I Nakayama, Masahiko Watanabe, Shigetsugu Hatakeyama
ABSTRACT
Increasing evidence shows that immune-mediated mechanisms may contribute to the pathogenesis of central nervous system disorders including cerebellar ataxias, as indicated by the aberrant production of neuronal surface antibodies. We previously reported a patient with cerebellar ataxia associated with production of a new anti-neuronal antibody, anti-seizure-related 6 homolog like 2 (Sez6l2). Sez6l2 is a type 1 membrane protein that is highly expressed in the hippocampus and cerebellar cortex and mice lacking Sez6l2 protein family members develop ataxia. Here we used a proteomics-based approach to show that serum derived from this patient recognizes the extracellular domain of Sez6l2 and that Sez6l2 protein binds to both adducin (ADD) and glutamate receptor 1 (GluR1). Our results indicate that Sez6l2 is one of the auxiliary subunits of the AMPA receptor and acts as a scaffolding protein to link GluR1 to ADD. Furthermore, Sez6l2 overexpression upregulates ADD phosphorylation, whereas siRNA-mediated downregulation of Sez612 prevents ADD phosphorylation, suggesting that Sez6l2 modulates AMPA-ADD signal transduction.
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension