Merck
  • Hsa_circ_0045714 regulates chondrocyte proliferation, apoptosis and extracellular matrix synthesis by promoting the expression of miR-193b target gene IGF1R.
Hsa_circ_0045714 regulates chondrocyte proliferation, apoptosis and extracellular matrix synthesis by promoting the expression of miR-193b target gene IGF1R.
Human cell (2017-08-11)
Bao-Feng Li, Ying Zhang, Jin Xiao, Fei Wang, Mei Li, Xiao-Ze Guo, Hui-Bin Xie, Hong Xia, Bei Chen
ABSTRACT
In recent years, some studies have been made on the effects of circular RNA (circRNA) in osteoarthritis (OA) and so on; however, its mechanisms remain to be further explored. Studies have shown that tumor necrosis factor-alpha can inhibit hsa_circ_0045714 expression in chondrocytes so as to upregulate miR-193b expression. Dual-luciferase reporter assay showed that insulin-like growth factor 1 receptor (IGF1R) is a key target gene of miR-193b. Hsa_circ_0045714 over-expression does not influence miR-193b expression, but can inhibit its transcriptional activity, thereby upregulating IGF1R expression. Hsa_circ_0045714 can promote the expression of type II collagen and aggrecan, and upregulate chondrocyte proliferation, while its linear sequences cannot. IGF1R has similar function, while miR-193b can inhibit the expression of type II collagen and aggrecan, and downregulate chondrocyte proliferation but enhance their apoptosis. IGF1R overexpression can reverse the effect of miR-193b, while miR-193b mimics or IGF1R siRNA can inhibit the function of hsa_circ_0045714. Therefore, hsa_circ_0045714 can regulate extracellular matrix synthesis as well as proliferation and apoptosis of chondrocytes by promoting the expression of miR-193b target gene IGF1R. The findings will provide new proofs for studies on the applications of circRNA in OA and other orthopedic diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human IGF1R