Merck
  • BRD4 regulates adiponectin gene induction by recruiting the P-TEFb complex to the transcribed region of the gene.
BRD4 regulates adiponectin gene induction by recruiting the P-TEFb complex to the transcribed region of the gene.
Scientific reports (2017-09-22)
Naoko Sakurai, Yuko Inamochi, Takuya Inoue, Natsuyo Hariya, Musashi Kawamura, Masami Yamada, Anup Dey, Akira Nishiyama, Takeo Kubota, Keiko Ozato, Toshinao Goda, Kazuki Mochizuki
ABSTRACT
We previously reported that induction of the adipocyte-specific gene adiponectin (Adipoq) during 3T3-L1 adipocyte differentiation is closely associated with epigenetic memory histone H3 acetylation on the transcribed region of the gene. We used 3T3-L1 adipocytes and Brd4 heterozygous mice to investigate whether the induction of Adipoq during adipocyte differentiation is regulated by histone acetylation and the binding protein bromodomain containing 4 (BRD4) on the transcribed region. Depletion of BRD4 by shRNA and inhibition by (+)-JQ1, an inhibitor of BET family proteins including BRD4, reduced Adipoq expression and lipid droplet accumulation in 3T3-L1 adipocytes. Additionally, the depletion and inhibition of BRD4 reduced the expression of many insulin sensitivity-related genes, including genes related to lipid droplet accumulation in adipocytes. BRD4 depletion reduced P-TEFb recruitment and histone acetylation on the transcribed region of the Adipoq gene. The expression levels of Adipoq and fatty acid synthesis-related genes and the circulating ADIPOQ protein level were lower in Brd4 heterozygous mice than in wild-type mice at 21 days after birth. These findings indicate that BRD4 regulates the Adipoq gene by recruiting P-TEFb onto acetylated histones in the transcribed region of the gene and regulates adipocyte differentiation by regulating the expression of genes related to insulin sensitivity.
Sigma-Aldrich
Triton X-100, laboratory grade