- Heterogeneous resistance to quizartinib in acute myeloid leukemia revealed by single-cell analysis.
Heterogeneous resistance to quizartinib in acute myeloid leukemia revealed by single-cell analysis.
Blood (2017-05-12)
Catherine C Smith, Amy Paguirigan, Grace R Jeschke, Kimberly C Lin, Evan Massi, Theodore Tarver, Chen-Shan Chin, Saurabh Asthana, Adam Olshen, Kevin J Travers, Susana Wang, Mark J Levis, Alexander E Perl, Jerald P Radich, Neil P Shah
PMID28490572
ABSTRACT
Genomic studies have revealed significant branching heterogeneity in cancer. Studies of resistance to tyrosine kinase inhibitor therapy have not fully reflected this heterogeneity because resistance in individual patients has been ascribed to largely mutually exclusive on-target or off-target mechanisms in which tumors either retain dependency on the target oncogene or subvert it through a parallel pathway. Using targeted sequencing from single cells and colonies from patient samples, we demonstrate tremendous clonal diversity in the majority of acute myeloid leukemia (AML) patients with activating
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.