Merck
IL-24 Promotes
Journal of immunology (Baltimore, Md. : 1950) (2017-03-24)
Bing X Ross, Nan Gao, Xinhan Cui, Theodore J Standiford, Jianjiang Xu, Fu-Shin X Yu
ABSTRACT
The aim of this study was to elucidate the expression and functions of IL-24 in C57BL/6 mouse corneas in response to
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture