Merck
  • miR-194 functions as a novel modulator of cellular senescence in mouse embryonic fibroblasts.
miR-194 functions as a novel modulator of cellular senescence in mouse embryonic fibroblasts.
Cell biology international (2016-12-17)
Shun Xu, Bing Zhang, Yanmei Zhu, Haijiao Huang, Wenping Yang, Haiyong Huang, Hui-Ling Zheng, Xinguang Liu
ABSTRACT
MicroRNA-194 (miR-194), a typical p53 responsive miRNA, serves as a tumor suppressor similar as p53, and has been demonstrated to play an anti-proliferation role in various human cancers. In spite of the pivotal role of p53 during aging process, the knowledge of miR-194's contribution to cellular senescence is limited. We herein sought to explore the role of miR-194 in the replicative senescence and stress-induced senescence of mouse embryonic fibroblasts. Our results unraveled that, compared to young cells, miR-194 is highly expressed in senescent cells, and extra expression of miR-194 significantly triggers the replicative senescence of MEFs and H
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.5 mL
Please contact Customer Service for Availability
$1,940.00