Merck
  • Loss of Sirt1 function improves intestinal anti-bacterial defense and protects from colitis-induced colorectal cancer.
Loss of Sirt1 function improves intestinal anti-bacterial defense and protects from colitis-induced colorectal cancer.
PloS one (2014-07-12)
Giuseppe Lo Sasso, Dongryeol Ryu, Laurent Mouchiroud, Samodha C Fernando, Christopher L Anderson, Elena Katsyuba, Alessandra Piersigilli, Michael O Hottiger, Kristina Schoonjans, Johan Auwerx
ABSTRACT
Dysfunction of Paneth and goblet cells in the intestine contributes to inflammatory bowel disease (IBD) and colitis-associated colorectal cancer (CAC). Here, we report a role for the NAD+-dependent histone deacetylase SIRT1 in the control of anti-bacterial defense. Mice with an intestinal specific Sirt1 deficiency (Sirt1int-/-) have more Paneth and goblet cells with a consequent rearrangement of the gut microbiota. From a mechanistic point of view, the effects on mouse intestinal cell maturation are mediated by SIRT1-dependent changes in the acetylation status of SPDEF, a master regulator of Paneth and goblet cells. Our results suggest that targeting SIRT1 may be of interest in the management of IBD and CAC.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution