Merck
  • Coxsackievirus B1 reveals strain specific differences in plasmacytoid dendritic cell mediated immunogenicity.
Coxsackievirus B1 reveals strain specific differences in plasmacytoid dendritic cell mediated immunogenicity.
Journal of medical virology (2014-03-13)
Sanna Hämäläinen, Noora Nurminen, Helena Ahlfors, Sami Oikarinen, Amir-Babak Sioofy-Khojine, Gun Frisk, M Steven Oberste, Riitta Lahesmaa, Marko Pesu, Heikki Hyöty
ABSTRACT
Enterovirus infections are usually mild but can also cause severe illnesses and play a role in chronic diseases, such as cardiomyopathies and type 1 diabetes. Host response to the invading virus can markedly modulate the course of the infection, and this response varies between individuals due to the polymorphism of immune response genes. However, it is currently not known if virus strains also differ in their ability to stimulate the host immune system. Coxsackievirus B1 (CBV1) causes severe epidemics in young infants and it has recently been connected with type 1 diabetes in seroepidemiological studies. This study evaluated the ability of different field isolates of CBV1 to induce innate immune responses in PBMCs. CBV1 strains differed markedly in their capacity to induce innate immune responses. Out of the 18 tested CBV1 strains two induced exceptionally strong alpha interferon (IFN-α) response in PBMC cultures. The responding cell type was found to be the plasmacytoid dendritic cell. Such a strong innate immune response was accompanied by an up-regulation of several other immune response genes and secretion of cytokines, which modulate inflammation, and adaptive immune responses. These results suggest that enterovirus-induced immune activation depends on the virus strain. It is possible that the immunotype of the virus modulates the course of the infection and plays a role in the pathogenesis of chronic immune-mediated enterovirus diseases.
Sigma-Aldrich
Rat IL-10 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-10 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-10 ELISA Kit