Merck
  • Using a new Lrig1 reporter mouse to assess differences between two Lrig1 antibodies in the intestine.
Using a new Lrig1 reporter mouse to assess differences between two Lrig1 antibodies in the intestine.
Stem cell research (2014-12-03)
Emily J Poulin, Anne E Powell, Yang Wang, Yina Li, Jeffrey L Franklin, Robert J Coffey
ABSTRACT
Lrig1 is an intestinal stem cell marker important for epithelial homeostasis. However, the position of the Lrig1(+) population in the intestinal crypt has been debated, largely due to discrepant staining patterns using two Lrig1 antibodies. Here, we set out to decipher the differences between these Lrig1 antibodies to clarify their use for Lrig1-related studies. We confirmed that the commercially available Lrig1-R&D antibody stained the bottom third of the colonic crypt, whereas an independently generated Lrig1-VU antibody recognized a subset of anti-Lrig1-R&D(+) cells. Biochemically, we found that anti-Lrig1-VU recognized a non-glycosylated form of Lrig1; in contrast, anti-Lrig1-R&D recognized both glycosylated and non-glycosylated forms of Lrig1. In addition, we generated a reporter mouse (Lrig1-Apple) as an independent readout of Lrig1 transcriptional activity. Flow cytometry of isolated colonic epithelial cells from Lrig1-Apple mice demonstrated anti-Lrig1-R&D recognized mostly RFP-hi cells, while anti-Lrig1-VU recognized cells that were largely RFP-mid. Of note, by qRT-PCR, Lgr5 was expressed in the RFP-hi population, but not in the RFP-mid population. We conclude that anti-Lrig1-R&D appears to recognize all Lrig1(+) cells, while anti-Lrig1-VU recognizes a subpopulation of Lrig1(+) cells.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride, AR, ≥99.9%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, ≥99%, AR grade
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Supelco
Digoxigenin, analytical standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets