Merck
  • Gp120 in the pathogenesis of human immunodeficiency virus-associated pain.
Gp120 in the pathogenesis of human immunodeficiency virus-associated pain.
Annals of neurology (2014-03-19)
Su-Bo Yuan, Yuqiang Shi, Jinghong Chen, Xiangfu Zhou, Guangyu Li, Benjamin B Gelman, Joshua G Lisinicchia, Susan M Carlton, Monique R Ferguson, Alai Tan, Sushil K Sarna, Shao-Jun Tang
ABSTRACT
Chronic pain is a common neurological comorbidity of human immunodeficiency virus (HIV)-1 infection, but the etiological cause remains elusive. The objective of this study was to identify the HIV-1 causal factor that critically contributes to the pathogenesis of HIV-associated pain. We first compared the levels of HIV-1 proteins in postmortem tissues of the spinal cord dorsal horn (SDH) from HIV-1/acquired immunodeficiency syndrome patients who developed chronic pain (pain-positive HIV-1 patients) and HIV-1 patients who did not develop chronic pain (pain-negative HIV-1 patients). Then we used the HIV-1 protein that was specifically increased in the pain-positive patients to generate mouse models. Finally, we performed comparative analyses on the pathological changes in the models and the HIV-1 patients. We found that HIV-1 gp120 was significantly higher in pain-positive HIV-1 patients (vs pain-negative HIV-1 patients). This finding suggested that gp120 was a potential causal factor of the HIV-associated pain. To test this hypothesis, we used a mouse model generated by intrathecal injection of gp120 and compared the pathologies of the model and the pain-positive human HIV-1 patients. The results showed that the mouse model and pain-positive human HIV-1 patients developed extensive similarities in their pathological phenotypes, including pain behaviors, peripheral neuropathy, glial reactivation, synapse degeneration, and aberrant activation of pain-related signaling pathways in the SDH. Our findings suggest that gp120 may critically contribute to the pathogenesis of HIV-associated pain.
Product Description
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Synapsin I Antibody, serum, Chemicon®