Merck
  • Rebound potentiation of inhibition in juvenile visual cortex requires vision-induced BDNF expression.
Rebound potentiation of inhibition in juvenile visual cortex requires vision-induced BDNF expression.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-08-08)
Ming Gao, Kristen R Maynard, Varun Chokshi, Lihua Song, Cara Jacobs, Hui Wang, Trinh Tran, Keri Martinowich, Hey-Kyoung Lee
ABSTRACT
The developmental increase in the strength of inhibitory synaptic circuits defines the time window of the critical period for plasticity in sensory cortices. Conceptually, plasticity of inhibitory synapses is an attractive mechanism to allow for homeostatic adaptation to the sensory environment. However, a brief duration of visual deprivation that causes maximal change in excitatory synapses produces minimal change in inhibitory synaptic transmission. Here we examined developmental and experience-dependent changes in inhibition by measuring miniature IPSCs (mIPSCs) in layer 2/3 pyramidal neurons of mouse visual cortex. During development from postnatal day 21 (P21) to P35, GABAA receptor function changed from fewer higher-conductance channels to more numerous lower-conductance channels without altering the average mIPSC amplitude. Although a week of visual deprivation did not alter the average mIPSC amplitude, a subsequent 2 h exposure to light produced a rapid rebound potentiation. This form of plasticity is restricted to a critical period before the developmental change in GABAergic synaptic properties is completed, and hence is absent by P35. Visual experience-dependent rebound potentiation of mIPSCs is accompanied by an increase in the open channel number and requires activity-dependent transcription of brain-derived neurotrophic factor (BDNF). Mice lacking BDNF transcription through promoter IV did not show developmental changes in inhibition and lacked rebound potentiation. Our results suggest that sensory experience may have distinct functional consequences in normal versus deprived sensory cortices, and that experience-dependent BDNF expression controls the plasticity of inhibitory synaptic transmission particularly when recovering vision during the critical period.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
HEPES, Vetec, reagent grade, 99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%