Merck
  • Genome-wide modulation of gene transcription in ovarian carcinoma cells by a new mithramycin analogue.
Genome-wide modulation of gene transcription in ovarian carcinoma cells by a new mithramycin analogue.
PloS one (2014-08-12)
Carolina Vizcaíno, Luz-Elena Núñez, Francisco Morís, José Portugal
ABSTRACT
Ovarian cancer has a poor prognosis due to intrinsic or acquired resistance to some cytotoxic drugs, raising the interest in new DNA-binding agents such as mithramycin analogues as potential chemotherapeutic agents in gynecological cancer. Using a genome-wide approach, we have analyzed gene expression in A2780 human ovarian carcinoma cells treated with the novel mithramycin analogue DIG-MSK (demycarosyl-3D-β-D-digitoxosyl-mithramycin SK) that binds to C+G-rich DNA sequences. Nanomolar concentrations of DIG-MSK abrogated the expression of genes involved in a variety of cell processes including transcription regulation and tumor development, which resulted in cell death. Some of those genes have been associated with cell proliferation and poor prognosis in ovarian cancer. Sp1 transcription factor regulated most of the genes that were down-regulated by the drug, as well as the up-regulation of other genes mainly involved in response to cell stress. The effect of DIG-MSK in the control of gene expression by other transcription factors was also explored. Some of them, such as CREB, E2F and EGR1, also recognize C/G-rich regions in gene promoters, which encompass potential DIG-MSK binding sites. DIG-MSK affected several biological processes and molecular functions related to transcription and its cellular regulation in A2780 cells, including transcription factor activity. This new compound might be a promising drug for the treatment of ovarian cancer.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Propidium iodide solution