Merck
  • Effects of combined administration of rapamycin, tolvaptan, and AEZ-131 on the progression of polycystic disease in PCK rats.
Effects of combined administration of rapamycin, tolvaptan, and AEZ-131 on the progression of polycystic disease in PCK rats.
American journal of physiology. Renal physiology (2014-03-22)
Massimo Sabbatini, Luigi Russo, Francesco Cappellaio, Giancarlo Troncone, Claudio Bellevicine, Valentina De Falco, Preziosa Buonocore, Eleonora Riccio, Vincenzo Bisesti, Stefano Federico, Antonio Pisani
ABSTRACT
Both experimental and clinical studies have suggested that any potential treatment of polycystic kidney disease (PKD) should start early and last for a long time to be effective, with unavoidable side reactions and considerable costs. The aim of the present study was to test how low doses of rapamycin (RAPA; 0.15 mg/kg ip for 4 days/wk), tolvaptan (TOLV; 0.005% in diet), or AEZ-131 (AEZ; a novel ERK inhibitor, 30 mg/kg for 3 days/wk by gavage), alone and in association, affect the progression of polycystic renal disease in PCK rats. Rats were treated for 8 wk starting at 4-6 wk of age. The efficacy of low doses of such drugs in inhibiting their respective targets was confirmed by immunoblot experiments. Compared with rats in the control (CON) group, RAPA treatment caused a significant reduction in cyst volume density (CVD; -19% vs. the CON group) and was numerically similar to that in TOLV-treated rats (-18%, not significiant), whereas AEZ treatment was not effective. RAPA + TOLV treatment resulted in a significantly lower CVD (-49% vs. the CON group) and was associated with a striking decrease in cAMP response element-binding protein phosphorylation, and similar data were detected in RAPA + AEZ-treated rats (-42%), whereas TOLV + AEZ treatment had virtually no effect. RAPA administration significantly lessened body weight gain, whereas TOLV administration resulted a mild increase in diuresis and a significant increase in cAMP urinary excretion. Histological data of tubular proliferation were in full agreement with CVD data. In conclusion, this study demonstrates that the association of low doses of RAPA, TOLV, and AEZ slows the progression of PKD with limited side effects, suggesting the use of combined therapies also in clinical trials.
Sigma-Aldrich
Anti-PCNA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution