Merck
  • Efficacy of planar chromatography coupled to (tandem) mass spectrometry for employment in trace analysis.
Efficacy of planar chromatography coupled to (tandem) mass spectrometry for employment in trace analysis.
Journal of chromatography. A (2006-07-28)
Ute Jautz, Gertrud Morlock
ABSTRACT
HPTLC/MS by a plunger-based extraction device was shown to be an appropriate technique for quantitative planar chromatography, even in trace analysis. Reproducible extraction from silica gel phases in the lower-pg range distinguishes this technique from other approaches. Repeatability of the MS signal showed a mean RSD of 12.5% for the example of Harman, a heterocyclic aromatic amine. Analytical response within a plate and over various plates/days showed determination coefficients of 0.9915 and 0.9488, respectively. Limit of detection (LOD) by a single quadrupole was better than 40 pg and limit of quantitation (LOQ) by a tandem mass spectrometer better than 20 pg. LOQ/LOD obtained were of similar magnitude as reported for HPLC/MS methods, however, this order of sensitivity was shown for the first time in the field of HPTLC/electrospray ionization MS.
Sigma-Aldrich
Harmane, 98%