Merck
  • Development of a manual method for the determination of mineral oil in foods and paperboard.
Development of a manual method for the determination of mineral oil in foods and paperboard.
Journal of chromatography. A (2012-12-12)
Katell Fiselier, Florian Grundböck, Karsten Schön, Oliver Kappenstein, Karla Pfaff, Christoph Hutzler, Andreas Luch, Koni Grob
ABSTRACT
So far the majority of the measurements of mineral oil saturated hydrocarbons (MOSH) and mineral oil aromatic hydrocarbons (MOAH) were obtained from on-line high performance liquid chromatography-gas chromatography-flame ionization detection (on-line HPLC-GC-FID). Since this technique is not available in many laboratories, an alternative method with more easily available tools has been developed. Preseparation on a small conventional liquid chromatographic column was optimized to achieve robust separation between the MOSH and the MOAH, but also to keep out the wax esters from the MOAH fraction. This was achieved by mixing a small portion of silica gel with silver nitrate into highly activated silica gel and by adding toluene into the eluent for the MOAH. Toluene was also added to the MOSH fraction to facilitate reconcentration and to serve as a keeper preventing loss of volatiles during solvent evaporation. A 50 μl volume was injected on-column into GC-FID to achieve a detection limit for MOSH and MOAH below 1 mg/kg in most foods.
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O