Merck
  • Structure-activity relationship of novel menaquinone-4 analogues: modification of the side chain affects their biological activities.
Structure-activity relationship of novel menaquinone-4 analogues: modification of the side chain affects their biological activities.
Journal of medicinal chemistry (2012-01-19)
Yoshitomo Suhara, Norika Hanada, Takashi Okitsu, Miho Sakai, Masato Watanabe, Kimie Nakagawa, Akimori Wada, Kazuyoshi Takeda, Kazuhiko Takahashi, Hiroaki Tokiwa, Toshio Okano
ABSTRACT
We synthesized new vitamin K analogues with demethylation or reduction of the double bonds of the side chain of menaquinone-4 (MK-4) and evaluated their SXR-mediated transcriptional activity as well as the extent of their conversion to MK-4. The results indicated that the analogue with the methyl group deleted at the 7' site of the side chain part affected conversion activity to MK-4. In contrast, a decrease in the number of the double bonds in the side chain moiety appeared to decrease the SXR-mediated transcriptional activity.
Product Description
Supelco
Menaquinone (K2), analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Vitamin K2