Merck
  • Imatinib attenuates reperfusion injury in a rat model of acute myocardial infarction.
Imatinib attenuates reperfusion injury in a rat model of acute myocardial infarction.
Basic research in cardiology (2023-01-14)
Lara S F Konijnenberg, Tom T J Luiken, Andor Veltien, Laween Uthman, Carolien T A Kuster, Laura Rodwell, Guus A de Waard, Mariska Kea-Te Lindert, Anat Akiva, Dick H J Thijssen, Robin Nijveldt, Niels van Royen
ABSTRACT
Following an acute myocardial infarction, reperfusion of an occluded coronary artery is often accompanied by microvascular injury, leading to worse long-term prognosis. Experimental studies have revealed the potential of tyrosine-kinase inhibitor imatinib to reduce vascular leakage in various organs. Here, we examined the potential of imatinib to attenuate microvascular injury in a rat model of myocardial reperfusion injury. Isolated male Wistar rat hearts (n = 20) in a Langendorff system and male Wistar rats (n = 37) in an in vivo model were randomly assigned to imatinib or placebo and subjected to ischaemia and reperfusion. Evans-blue/Thioflavin-S/TTC staining and Cardiac Magnetic Resonance Imaging were performed to assess the extent of reperfusion injury. Subsequently, in vivo hearts were perfused ex vivo with a vascular leakage tracer and fluorescence and electron microscopy were performed. In isolated rat hearts, imatinib reduced global infarct size, improved end-diastolic pressure, and improved rate pressure product recovery compared to placebo. In vivo, imatinib reduced no-reflow and infarct size with no difference between imatinib and placebo for global cardiac function. In addition, imatinib showed lower vascular resistance, higher coronary flow, and less microvascular leakage in the affected myocardium. At the ultrastructural level, imatinib showed higher preserved microvascular integrity compared to placebo. We provide evidence that low-dose imatinib can reduce microvascular injury and accompanying myocardial infarct size in a rat model of acute myocardial infarction. These data warrant future work to examine the potential of imatinib to reduce reperfusion injury in patients with acute myocardial infarction.
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), FITC conjugate, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Imatinib mesylate, ≥98% (HPLC)