Merck
  • A new fluorescent quenching method for the determination of phosphoproteins by using calconcarboxylic acid.
A new fluorescent quenching method for the determination of phosphoproteins by using calconcarboxylic acid.
Electrophoresis (2014-12-30)
Zhongxin Zhu, Xinliang Zhu, Jiayi Shen, Ayi Zhou, Maowei Ni, Litai Jin, Weitao Cong
ABSTRACT
A fluorescent quenching detection method for phosphoproteins in SDS-PAGE by using calconcarboxylic acid (CCA) was described. In this method, the fluorescence intensity of CCA was greatly increased with the presence of Al(3+) in the gel background, while in zones where phosphoproteins are located this intensity was absent because of fluorescence quenching phenomenon through the formation of CCA-Al(3+) -phosphoprotein appended complex. Approximately 4-8 ng of phosphoproteins can be selectively detected within 1 h (1D SDS-PAGE), which is similar to that of the most commonly used Pro-Q Diamond stain. The specificity of this novel technique for phosphoproteins was confirmed by dephosphorylation, Western blot, and LC-MS/MS analysis, respectively. Furthermore, to better understand the newly developed method, the detection mechanism of CCA stain was explored by fluorescent spectrometry. According to the results, it is believed that CCA stain may provide a new choice for selective, economical, MS compatible, and convenient visualization of gel-separated phosphoproteins.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Pricing and availability is not currently available.