Merck
  • A phosphorylation-regulated eIF3d translation switch mediates cellular adaptation to metabolic stress.
A phosphorylation-regulated eIF3d translation switch mediates cellular adaptation to metabolic stress.
Science (New York, N.Y.) (2020-11-14)
Adam M Lamper, Rebecca H Fleming, Kayla M Ladd, Amy S Y Lee
ABSTRACT
Shutoff of global protein synthesis is a conserved response to cellular stresses. This general phenomenon is accompanied by the induction of distinct gene programs tailored to each stress. Although the mechanisms driving repression of general protein synthesis are well characterized, how cells reprogram the translation machinery for selective gene expression remains poorly understood. Here, we found that the noncanonical 5' cap-binding protein eIF3d was activated in response to metabolic stress in human cells. Activation required reduced CK2-mediated phosphorylation near the eIF3d cap-binding pocket. eIF3d controls a gene program enriched in factors important for glucose homeostasis, including members of the mammalian target of rapamycin (mTOR) pathway. eIF3d-directed translation adaptation was essential for cell survival during chronic glucose deprivation. Thus, this mechanism of translation reprogramming regulates the cellular response to metabolic stress.