Merck
  • Suppression of DNA Double-Strand Break Formation by DNA Polymerase β in Active DNA Demethylation Is Required for Development of Hippocampal Pyramidal Neurons.
Suppression of DNA Double-Strand Break Formation by DNA Polymerase β in Active DNA Demethylation Is Required for Development of Hippocampal Pyramidal Neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2020-10-23)
Akiko Uyeda, Kohei Onishi, Teruyoshi Hirayama, Satoko Hattori, Tsuyoshi Miyakawa, Takeshi Yagi, Nobuhiko Yamamoto, Noriyuki Sugo
ABSTRACT
Genome stability is essential for brain development and function, as de novo mutations during neuronal development cause psychiatric disorders. However, the contribution of DNA repair to genome stability in neurons remains elusive. Here, we demonstrate that the base excision repair protein DNA polymerase β (Polβ) is involved in hippocampal pyramidal neuron differentiation via a TET-mediated active DNA demethylation during early postnatal stages using Nex-Cre/Polβ fl/fl mice of either sex, in which forebrain postmitotic excitatory neurons lack Polβ expression. Polβ deficiency induced extensive DNA double-strand breaks (DSBs) in hippocampal pyramidal neurons, but not dentate gyrus granule cells, and to a lesser extent in neocortical neurons, during a period in which decreased levels of 5-methylcytosine and 5-hydroxymethylcytosine were observed in genomic DNA. Inhibition of the hydroxylation of 5-methylcytosine by expression of microRNAs miR-29a/b-1 diminished DSB formation. Conversely, its induction by TET1 catalytic domain overexpression increased DSBs in neocortical neurons. Furthermore, the damaged hippocampal neurons exhibited aberrant neuronal gene expression profiles and dendrite formation, but not apoptosis. Comprehensive behavioral analyses revealed impaired spatial reference memory and contextual fear memory in adulthood. Thus, Polβ maintains genome stability in the active DNA demethylation that occurs during early postnatal neuronal development, thereby contributing to differentiation and subsequent learning and memory.SIGNIFICANCE STATEMENT Increasing evidence suggests that de novo mutations during neuronal development cause psychiatric disorders. However, strikingly little is known about how DNA repair is involved in neuronal differentiation. We found that Polβ, a component of base excision repair, is required for differentiation of hippocampal pyramidal neurons in mice. Polβ deficiency transiently led to increased DNA double-strand breaks, but not apoptosis, in early postnatal hippocampal pyramidal neurons. This aberrant double-strand break formation was attributed to active DNA demethylation as an epigenetic regulation. Furthermore, the damaged neurons exhibited aberrant gene expression profiles and dendrite formation, resulting in impaired learning and memory in adulthood. Thus, these findings provide new insight into the contribution of DNA repair to the neuronal genome in early brain development.
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, Cy3 conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
2-Phospho-L-ascorbic acid trisodium salt, ≥95.0% (HPLC)
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Methyl methanesulfonate, 99%