Merck
  • ELL-associated factors EAF1/2 negatively regulate HIV-1 transcription through inhibition of Super Elongation Complex formation.
ELL-associated factors EAF1/2 negatively regulate HIV-1 transcription through inhibition of Super Elongation Complex formation.
Biochimica et biophysica acta. Gene regulatory mechanisms (2020-02-23)
Rongdiao Liu, Chunjing Chen, Yun Li, Qingqing Huang, Yuhua Xue
ABSTRACT
The ELL (ELL1 and ELL2)-containing Super Elongation Complex (SEC) is required for efficient HIV-1 transactivation by the viral-encoded Tat protein. EAF1 and EAF2 are ELL-associated factors and considered as positive regulators of ELL. However, their role in HIV-1 transcriptional control is unknown. In this study, we show that EAF1/2 inhibit the SEC-dependent and Tat-activated HIV-1 transcription. EAF1/2 are found to interact with the SEC components in an ELL1/2-dependent manner. Surprisingly, the depletion of EAF1/2 increases the SEC formation and occupancy on the HIV-1 proviral DNA, thereby stimulating Tat transactivation of HIV-1. Although EAF1/2 interact with members of the SEC in a ELL-dependent manner, this interaction competes with the binding of the scaffolding subunit AFF1 with ELL, thus reducing the SEC formation. Together, these data reveal how EAF1/2 regulate the SEC formation to control HIV-1 transcription.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)