Merck
  • Treatment-Induced Tumor Dormancy through YAP-Mediated Transcriptional Reprogramming of the Apoptotic Pathway.
Treatment-Induced Tumor Dormancy through YAP-Mediated Transcriptional Reprogramming of the Apoptotic Pathway.
Cancer cell (2020-01-15)
Kari J Kurppa, Yao Liu, Ciric To, Tinghu Zhang, Mengyang Fan, Amir Vajdi, Erik H Knelson, Yingtian Xie, Klothilda Lim, Paloma Cejas, Andrew Portell, Patrick H Lizotte, Scott B Ficarro, Shuai Li, Ting Chen, Heidi M Haikala, Haiyun Wang, Magda Bahcall, Yang Gao, Sophia Shalhout, Steffen Boettcher, Bo Hee Shin, Tran Thai, Margaret K Wilkens, Michelle L Tillgren, Mierzhati Mushajiang, Man Xu, Jihyun Choi, Arrien A Bertram, Benjamin L Ebert, Rameen Beroukhim, Pratiti Bandopadhayay, Mark M Awad, Prafulla C Gokhale, Paul T Kirschmeier, Jarrod A Marto, Fernando D Camargo, Rizwan Haq, Cloud P Paweletz, Kwok-Kin Wong, David A Barbie, Henry W Long, Nathanael S Gray, Pasi A Jänne
ABSTRACT
Eradicating tumor dormancy that develops following epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKI) treatment of EGFR-mutant non-small cell lung cancer, is an attractive therapeutic strategy but the mechanisms governing this process are poorly understood. Blockade of ERK1/2 reactivation following EGFR TKI treatment by combined EGFR/MEK inhibition uncovers cells that survive by entering a senescence-like dormant state characterized by high YAP/TEAD activity. YAP/TEAD engage the epithelial-to-mesenchymal transition transcription factor SLUG to directly repress pro-apoptotic BMF, limiting drug-induced apoptosis. Pharmacological co-inhibition of YAP and TEAD, or genetic deletion of YAP1, all deplete dormant cells by enhancing EGFR/MEK inhibition-induced apoptosis. Enhancing the initial efficacy of targeted therapies could ultimately lead to prolonged treatment responses in cancer patients.
Sigma-Aldrich
Di(N-succinimidyl) glutarate, ≥97.0% (CHN)
Sigma-Aldrich
MISSION® esiRNA, targeting human SNAI2
Sigma-Aldrich
MISSION® esiRNA, targeting human BMF
Sigma-Aldrich
WZ4002, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human YAP1