Merck
  • WWP2 regulates proliferation of gastric cancer cells in a PTEN-dependent manner.
WWP2 regulates proliferation of gastric cancer cells in a PTEN-dependent manner.
Biochemical and biophysical research communications (2019-11-05)
Ke Wang, Jun Liu, Xinhui Zhao, Haitong Li, Guangwei Luo, Yanping Yu, Yuan Guo, Long Zhang, Jun Zhu, Shuai Wang, Wei Hua, Angang Yang, Rui Zhang, Jipeng Li
ABSTRACT
WW domain containing E3 Ub-protein ligase 2 (WWP2) plays an important role in tumor progression as an E3 ligase of PTEN. Here, we investigated the role of WWP2 in gastric cancer (GC). We found that WWP2 is overexpressed in GC tissues, which is closely related to poor prognosis of GC patients. Using a WWP2-shRNA lentivirus expressing system, we established WWP2 stable-knockdown GC cell lines and found that knockdown of WWP2 inhibits the proliferation of GC cells both in vitro and in vivo. Also, WWP2 silencing induced the up-regulation of PTEN protein level and down-regulation of AKT phosphorylation level. We further investigated the role of PTEN in this regulating process by performing rescue assay and found that PTEN is essential for WWP2-mediated regulation of GC cells proliferation. Taken together, our results demonstrated that WWP2 promotes proliferation of GC cells by downregulating PTEN, which may provide new therapeutic targets for GC.
Sigma-Aldrich
MISSION® esiRNA, targeting human PTEN