Merck
  • Activation of Skeletal Muscle AMPK Promotes Glucose Disposal and Glucose Lowering in Non-human Primates and Mice.
Activation of Skeletal Muscle AMPK Promotes Glucose Disposal and Glucose Lowering in Non-human Primates and Mice.
Cell metabolism (2017-05-04)
Emily C Cokorinos, Jake Delmore, Allan R Reyes, Bina Albuquerque, Rasmus Kjøbsted, Nicolas O Jørgensen, Jean-Luc Tran, Aditi Jatkar, Katherine Cialdea, Ryan M Esquejo, John Meissen, Matthew F Calabrese, Jason Cordes, Robert Moccia, David Tess, Christopher T Salatto, Timothy M Coskran, Alan C Opsahl, Declan Flynn, Matthew Blatnik, Wenlin Li, Erick Kindt, Marc Foretz, Benoit Viollet, Jessica Ward, Ravi G Kurumbail, Amit S Kalgutkar, Jørgen F P Wojtaszewski, Kimberly O Cameron, Russell A Miller
ABSTRACT
The AMP-activated protein kinase (AMPK) is a potential therapeutic target for metabolic diseases based on its reported actions in the liver and skeletal muscle. We evaluated two distinct direct activators of AMPK: a non-selective activator of all AMPK complexes, PF-739, and an activator selective for AMPK β1-containing complexes, PF-249. In cells and animals, both compounds were effective at activating AMPK in hepatocytes, but only PF-739 was capable of activating AMPK in skeletal muscle. In diabetic mice, PF-739, but not PF-249, caused a rapid lowering of plasma glucose levels that was diminished in the absence of skeletal muscle, but not liver, AMPK heterotrimers and was the result of an increase in systemic glucose disposal with no impact on hepatic glucose production. Studies of PF-739 in cynomolgus monkeys confirmed translation of the glucose lowering and established activation of AMPK in skeletal muscle as a potential therapeutic approach to treat diabetic patients.
Sigma-Aldrich
CelLytic MEM Protein Extraction Kit