Merck
  • RbAp46/48(LIN-53) Is Required for Holocentromere Assembly in Caenorhabditis elegans.
RbAp46/48(LIN-53) Is Required for Holocentromere Assembly in Caenorhabditis elegans.
Cell reports (2016-02-26)
Bernard Chi Hang Lee, Zhongyang Lin, Karen Wing Yee Yuen
ABSTRACT
Centromeres, the specialized chromosomal regions for recruiting kinetochores and directing chromosome segregation, are epigenetically marked by a centromeric histone H3 variant, CENP-A. To maintain centromere identity through cell cycles, CENP-A diluted during DNA replication is replenished. The licensing factor M18BP1(KNL-2) is known to recruit CENP-A to holocentromeres. Here, we show that RbAp46/48(LIN-53), a conserved histone chaperone, is required for CENP-A(HCP-3) localization in holocentric Caenorhabditis elegans. Indeed, RbAp46/48(LIN-53) and CENP-A(HCP-3) localizations are interdependent. RbAp46/48(LIN-53) localizes to the centromere during metaphase in a CENP-A(HCP-3)- and M18BP1(KNL-2)-dependent manner, suggesting CENP-A(HCP-3) loading may occur before anaphase. RbAp46/48(LIN-53) does not function at the centromere through histone acetylation, H3K27 trimethylation, or its known chromatin-modifying complexes. RbAp46/48(LIN-53) may function independently to escort CENP-A(HCP-3) for holocentromere assembly but is dispensable for other kinetochore protein recruitment. Nonetheless, depletion of RbAp46/48(LIN-53) leads to anaphase bridges and chromosome missegregation. This study unravels the holocentromere assembly hierarchy and its conservation with monocentromeres.
Sigma-Aldrich
Anti-acetyl Histone H3 (Lys9) Antibody, from rabbit, purified by affinity chromatography