Merck
  • Identification of novel predictive factors for post surgical corneal haze.
Identification of novel predictive factors for post surgical corneal haze.
Scientific reports (2019-11-20)
Nimisha R Kumar, Pooja Khamar, Rohit Shetty, Ankit Sharma, Naren Shetty, Natasha Pahuja, Valsala Gopalakrishnan Abilash, Vishal Jhanji, Anuprita Ghosh, Rajiv R Mohan, Rajani Kanth Vangala, Arkasubhra Ghosh
ABSTRACT
Molecular factors altered in corneas that develop haze post refractive surgery have been described, but pre-existing factors that predispose clinically normal corneas to aberrant fibrosis post surgery and the role of the corneal epithelium remains unknown. We analyzed the global gene expression in epithelium collected intraoperatively from subjects undergoing photorefractive keratectomy. Subjects were grouped into those that developed haze 12 months post surgery (n = 6 eyes; haze predisposed) and those that did not develop haze in a similar follow up duration (n = 11 eyes; controls). Ontological analysis of 1100 upregulated and 1780 downregulated genes in the haze predisposed group revealed alterations in pathways associated with inflammation, wnt signaling, oxidative stress, nerve functions and extra cellular matrix remodeling. Novel factors such as PREX1, WNT3A, SOX17, GABRA1and PXDN were found to be significantly altered in haze predisposed subjects and those with active haze(n = 3), indicating their pro-fibrotic role. PREX1 was significantly upregulated in haze predisposed subjects. Ectopic expression of PREX1 in cultured human corneal epithelial cells enhanced their rate of wound healing while its ablation using shRNA reduced healing compared to matched controls. Recombinant TGFβ treatment in PREX1 overexpressing corneal cells led to enhanced αSMA expression and Vimentin phosphorylation while the converse was true for shPREX1 expressing cells. Our data identify a few novel factors in the corneal epithelium that may define a patient's risk to developing post refractive corneal haze.
Sigma-Aldrich
MISSION® esiRNA, targeting human PREX1