Merck
  • In Situ Formation of Ag Nanoparticles in Mesoporous TiO2 Films Decorated on Bamboo via Self-Sacrificing Reduction to Synthesize Nanocomposites with Efficient Antifungal Activity.
In Situ Formation of Ag Nanoparticles in Mesoporous TiO2 Films Decorated on Bamboo via Self-Sacrificing Reduction to Synthesize Nanocomposites with Efficient Antifungal Activity.
International journal of molecular sciences (2019-11-07)
Jingpeng Li, Minglei Su, Anke Wang, Zaixing Wu, Yuhe Chen, Daochun Qin, Zehui Jiang
ABSTRACT
We developed a novel green approach for the in situ fabrication of Ag NPs in mesoporous TiO2 films via the bamboo self-sacrificing reduction of Ag(NH3)2+ ions, which can inhibit fungal growth on the bamboo surface. Mesoporous anatase TiO2 (MT) films were first synthesized on bamboo via a hydrothermal method. Then, Ag NPs with a 5.3 nm mean diameter were incorporated into the pore channels of optimal MT/bamboo (MTB) samples at room temperature without the addition of reducing agents, such that the Ag NPs were almost entirely embedded into the MT films. Our analysis indicated that the solubilized lignin from bamboo, which is rich in oxygen-containing functional groups, serves as a green reductant for reducing the Ag(NH3)2+ ions to Ag NPs. Antifungal experiments with Trichoderma viride under dark conditions highlighted that the antifungal activity of the Ag/MT/bamboo samples were greater than those of naked bamboo, MTB, and Ag/bamboo, suggesting that these hybrid nanomaterials produce a synergistic antifungal effect that is unrelated to photoactivity. The inhibition of Penicillium citrinum effectively followed a similar trend. This newly developed bamboo protection method may provide a sustainable, eco-friendly, and efficient method for enhancing the antifungal characteristics of traditional bamboo, having the potential to prolong the service life of bamboo materials, particularly under dark conditions.
Sigma-Aldrich
Ammonium hexafluorotitanate, 99.99% trace metals basis