- Superstretchable Dynamic Polymer Networks.
Superstretchable Dynamic Polymer Networks.
Advanced materials (Deerfield Beach, Fla.) (2019-09-07)
Huan Zhang, Yanzhou Wu, Jinxia Yang, Dong Wang, Pengyun Yu, Chi To Lai, An-Chang Shi, Jianping Wang, Shuxun Cui, Junfeng Xiang, Ning Zhao, Jian Xu
PMID31490600
ABSTRACT
Superstretchable materials have many applications in advanced technological fields but are difficult to stretch to more than 1000× their original length. A superstretchable dynamic polymer network that can be stretched to 13 000× its original length is designed. It is revealed that superstretchability of the polymer network is derived from the synergistic effect of two different types of dynamic bonds, including a small number of strong dynamic imine bonds to maintain the network integrity during stretching and a large number of weak ionic hydrogen bonds to dissipate energy. This approach provides new insights into the design of superstretchable polymers.