Merck
  • TAMM41 is required for heart valve differentiation via regulation of PINK-PARK2 dependent mitophagy.
TAMM41 is required for heart valve differentiation via regulation of PINK-PARK2 dependent mitophagy.
Cell death and differentiation (2019-03-03)
Rui Meng Yang, Jiong Tao, Ming Zhan, Hao Yuan, Hai Hong Wang, Sai Juan Chen, Zhu Chen, Hugues de Thé, Jun Zhou, Ying Guo, Jun Zhu
ABSTRACT
TAMM41, located within the congenital heart diseases (CHD) sensitive region of 3p25 deletion syndrome, is a mitochondrial membrane maintenance protein critical for yeast survival, but its function in higher vertebrates remains unknown. Via in vivo zebrafish model, we found that tamm41 is highly expressed in the developing heart and deficiency of which led to heart valve abnormalities. Molecular mechanistic studies revealed that TAMM41 interacts and modulates the PINK1-PARK2 dependent mitophagy pathway, thereby implicating TAMM41 in heart valve development during zebrafish embryonic cardiogenesis. Furthermore, through screening of the congenital heart diseases (CHD) sensitive region of 3p25 deletion syndrome among 118 sporadic atrioventricular septal defect (AVSD) patients, we identified three cases carrying heterozygous pathogenic intronic variants of TAMM41. All three cases lacked normal full-length TAMM41 transcripts, most likely due to specific expression of the mutant allele. Collectively, our studies highlight essential roles for TAMM41-dependent mitophagy in development of the heart and provide novel insights into the etiology of AVSD.
Product Description
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks