Merck
  • Store-operated Ca2+ entry in sensory neurons: functional role and the effect of painful nerve injury.
Store-operated Ca2+ entry in sensory neurons: functional role and the effect of painful nerve injury.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2011-03-11)
Geza Gemes, Madhavi Latha Yadav Bangaru, Hsiang-En Wu, Qingbo Tang, Dorothee Weihrauch, Andrew S Koopmeiners, James M Cruikshank, Wai-Meng Kwok, Quinn H Hogan
ABSTRACT
Painful nerve injury disrupts levels of cytoplasmic and stored Ca(2+) in sensory neurons. Since influx of Ca(2+) may occur through store-operated Ca(2+) entry (SOCE) as well as voltage- and ligand-activated pathways, we sought confirmation of SOCE in sensory neurons from adult rats and examined whether dysfunction of SOCE is a possible pathogenic mechanism. Dorsal root ganglion neurons displayed a fall in resting cytoplasmic Ca(2+) concentration when bath Ca(2+) was withdrawn, and a subsequent elevation of cytoplasmic Ca(2+) concentration (40 ± 5 nm) when Ca(2+) was reintroduced, which was amplified by store depletion with thapsigargin (1 μm), and was significantly reduced by blockers of SOCE, but was unaffected by antagonists of voltage-gated membrane Ca(2+) channels. We identified the underlying inwardly rectifying Ca(2+)-dependent I(CRAC) (Ca(2+) release activated current), as well as a large thapsigargin-sensitive inward current activated by withdrawal of bath divalent cations, representing SOCE. Molecular components of SOCE, specifically STIM1 and Orai1, were confirmed in sensory neurons at both the transcript and protein levels. Axonal injury by spinal nerve ligation (SNL) elevated SOCE and I(CRAC). However, SOCE was comparable in injured and control neurons when stores were maximally depleted by thapsigargin, and STIM1 and Orai1 levels were not altered by SNL, showing that upregulation of SOCE after SNL is driven by store depletion. Blockade of SOCE increased neuronal excitability in control and injured neurons, whereas injured neurons showed particular dependence on SOCE for maintaining levels of cytoplasmic and stored Ca(2+), which indicates a compensatory role for SOCE after injury.
Sigma-Aldrich
8-(3-Chlorostyryl)caffeine, ≥98% (HPLC), solid
Sigma-Aldrich
7-Nitroindazole, ≥98%
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film