Merck
Mechanistic insight into the regulation of SQSTM1/p62.
Autophagy (2019-01-18)
Yi Zhang, Su Ran Mun, Juan F Linares, Christina G Towers, Andrew Thorburn, Maria T Diaz-Meco, Yong Tae Kwon, Tatiana G Kutateladze
ABSTRACT
SQSTM1/p62 facilitates responses to various cellular stresses and has been implicated in human diseases. This protein functions as a major cytoplasmic signaling hub and has multiple binding partners, including arginylated (Nt-R) proteins that are recognized by the ZZ domain of SQSTM1/p62 (SQSTM1/p62ZZ). We have determined the molecular mechanism of Nt-R recognition using a combination of biochemical and NMR approaches and obtained the crystal structure of SQSTM1/p62ZZ in complex with Nt-R. We found that binding of SQSTM1/p62ZZ to Nt-R induces SQSTM1/p62 puncta formation and macroautophagy/autophagy and identified a regulatory linker (RL) region of SQSTM1/p62 that associates with SQSTM1/p62ZZ in vitro. Our findings suggest a mechanism for SQSTM1/p62 autoregulation that can be essential in mediating autophagy.
Sigma-Aldrich
XIE62-1004, ≥98% (HPLC)