Merck
  • The transcriptional landscape of lncRNAs reveals the oncogenic function of LINC00511 in ER-negative breast cancer.
The transcriptional landscape of lncRNAs reveals the oncogenic function of LINC00511 in ER-negative breast cancer.
Cell death & disease (2019-08-10)
Jian Zhang, Shiyao Sui, Hao Wu, Jinfeng Zhang, Xingda Zhang, Shouping Xu, Da Pang
ABSTRACT
Advances in the molecular characteristics of cancers have facilitated the classification system from morphology to molecular characteristic-based subtypes. Cancer profiling has expanded in its focus from protein-coding genes to noncoding RNAs, with advances in the depth and quality of transcriptome sequencing. Here, we examined the profiles of long noncoding RNAs (lncRNAs) according to breast cancer subtype categories in The Cancer Genome Atlas (TCGA) database to identify a cohort of breast cancer- and oestrogen receptor (ER)-negative-associated lncRNAs. According to the prioritization of variation in ER-negative-associated lncRNAs, we identified and investigated the role of LINC00511 in breast cancer. We determined that high LINC00511 expression was an unfavourable prognostic factor for patients with breast cancer. Furthermore, LINC00511 promoted tumour growth by accelerating the G1/S transition and inhibiting apoptosis. At the transcriptional level, ER deficiency directly affected the expression of LINC00511 activated by transcription factor AP-2 (TFAP-2) in breast cancer cells. Moreover, mechanistic investigations demonstrated that ER-negative-associated LINC00511 interacted with enhancer of zeste homologue 2 (EZH2, the catalytic subunit of polycomb repressive complex 2, PRC2) and recruited PRC2 to mediate histone methylation, contributing to the repression of CDKN1B in the nucleus. This process resulted in altered ER-negative breast cancer cell biology. By highlighting the oncogenic function of LINC00511, we revealed the role of lncRNAs in regulating the network of cell cycle control in ER-negative breast cancer and suggested the exploitation of LINC00511 as an anticancer therapy in the future.
Sigma-Aldrich
Tamoxifen, ≥99%